Product Details

SNP ID
rs3768809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:210197404 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCATTTCTATTCATCATATATTTA[A/C]TTAGGTCAAATATCTACCCTTTAAT
Phenotype
MIM: 609576
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ACADL PubMed Links

Gene Details

Gene
ACADL
Gene Name
acyl-CoA dehydrogenase, long chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001608.3 Intron NP_001599.1
XM_005246517.4 Intron XP_005246574.1
XM_017003955.1 Intron XP_016859444.1

View Full Product Details