Product Details

SNP ID
rs3208356
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:129650552 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACACTTAAATATCCACACATTT[C/T]TTCATTTAAAAACATCACTTCTTTA
Phenotype
MIM: 616242
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMCC1 PubMed Links

Gene Details

Gene
TMCC1
Gene Name
transmembrane and coiled-coil domain family 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017395.3 3048 UTR 3 NP_001017395.2
NM_001128224.2 3048 UTR 3 NP_001121696.1
XM_006713542.3 3048 UTR 3 XP_006713605.1
XM_006713543.3 3048 UTR 3 XP_006713606.1
XM_006713544.1 3048 UTR 3 XP_006713607.1
XM_006713550.3 3048 UTR 3 XP_006713613.1
XM_006713552.3 3048 UTR 3 XP_006713615.1
XM_006713553.3 3048 UTR 3 XP_006713616.1
XM_011512572.2 3048 UTR 3 XP_011510874.1
XM_011512573.2 3048 UTR 3 XP_011510875.1
XM_011512576.2 3048 UTR 3 XP_011510878.1
XM_011512579.2 3048 UTR 3 XP_011510881.1
XM_011512580.2 3048 UTR 3 XP_011510882.1
XM_011512581.2 3048 UTR 3 XP_011510883.1
XM_011512582.1 3048 UTR 3 XP_011510884.1
XM_011512583.2 3048 UTR 3 XP_011510885.1
XM_011512584.2 3048 UTR 3 XP_011510886.1
XM_017005931.1 3048 UTR 3 XP_016861420.1
XM_017005932.1 3048 UTR 3 XP_016861421.1
XM_017005933.1 3048 UTR 3 XP_016861422.1
XM_017005934.1 3048 UTR 3 XP_016861423.1
XM_017005935.1 3048 UTR 3 XP_016861424.1
XM_017005936.1 3048 UTR 3 XP_016861425.1
XM_017005937.1 3048 UTR 3 XP_016861426.1
XM_017005938.1 3048 UTR 3 XP_016861427.1
XM_017005939.1 3048 UTR 3 XP_016861428.1
XM_017005940.1 3048 UTR 3 XP_016861429.1
XM_017005941.1 3048 UTR 3 XP_016861430.1

View Full Product Details