Product Details
- SNP ID
-
rs3770753
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:37348434 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGAATTTTTTATATCAGTGGCATC[C/G]GACACACAGAAGGAGTTCAGTAACT
- Phenotype
-
MIM: 607065
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
QPCT
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75626957] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- QPCT
- Gene Name
- glutaminyl-peptide cyclotransferase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_012413.3 |
|
Intron |
|
|
NP_036545.1 |
View Full Product Details