Product Details

SNP ID
rs3795442
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:225987843 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
AGCACATTTTCATTTTAATGACTTA[C/T]TGTGGAGATTAAACTGATTTGGCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SDE2 PubMed Links

Gene Details

Gene
SDE2
Gene Name
SDE2 telomere maintenance homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152608.3 Intron NP_689821.3

View Full Product Details