Product Details

SNP ID
rs3813485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:3269887 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTAGGAAGCGGTGGTGTCTGAGTG[C/T]GATACTTCCCTTACGAGGTTTGTTT
Phenotype
MIM: 611697
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSMG4 PubMed Links

Gene Details

Gene
PSMG4
Gene Name
proteasome assembly chaperone 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128591.1 5793 Intron NP_001122063.1
NM_001128592.1 5793 Intron NP_001122064.1
NM_001135750.1 5793 Intron NP_001129222.1
XM_011514591.2 5793 Intron XP_011512893.1
XM_011514593.2 5793 Intron XP_011512895.1
XM_011514594.2 5793 Intron XP_011512896.1
XM_011514595.2 5793 Intron XP_011512897.1
Gene
SLC22A23
Gene Name
solute carrier family 22 member 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286455.1 5793 UTR 3 NP_001273384.1
NM_001286456.1 5793 Intron NP_001273385.1
NM_015482.1 5793 UTR 3 NP_056297.1
NM_021945.6 5793 UTR 3 NP_068764.3
XM_011514801.2 5793 UTR 3 XP_011513103.1
XM_017011180.1 5793 UTR 3 XP_016866669.1
XM_017011181.1 5793 UTR 3 XP_016866670.1
XM_017011182.1 5793 UTR 3 XP_016866671.1
XM_017011183.1 5793 Intron XP_016866672.1
XM_017011184.1 5793 UTR 3 XP_016866673.1
XM_017011185.1 5793 Intron XP_016866674.1
XM_017011186.1 5793 UTR 3 XP_016866675.1

View Full Product Details