Product Details

SNP ID
rs3731918
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219550995 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGGTTAGGGCTGTCTTGGCGGCA[A/G]GAAGGTGGGGGTCAGCTAGGGGTGG
Phenotype
MIM: 610405 MIM: 610991
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHPF PubMed Links

Gene Details

Gene
CHPF
Gene Name
chondroitin polymerizing factor
There are no transcripts associated with this gene.

Gene
MIR3132
Gene Name
microRNA 3132
There are no transcripts associated with this gene.

Gene
OBSL1
Gene Name
obscurin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173408.1 6314 Intron NP_001166879.1
NM_001173431.1 6314 Intron NP_001166902.1
NM_015311.2 6314 Intron NP_056126.1
XM_005246424.4 6314 Intron XP_005246481.1
XM_005246427.4 6314 Intron XP_005246484.1
XM_011510857.2 6314 Intron XP_011509159.1
XM_011510863.2 6314 Intron XP_011509165.1
XM_011510864.2 6314 Intron XP_011509166.1
XM_011510865.2 6314 Intron XP_011509167.1
XM_011510866.2 6314 Intron XP_011509168.1
XM_017003696.1 6314 UTR 3 XP_016859185.1
XM_017003697.1 6314 Intron XP_016859186.1
XM_017003698.1 6314 Intron XP_016859187.1
XM_017003699.1 6314 Intron XP_016859188.1
XM_017003700.1 6314 Intron XP_016859189.1
Gene
TMEM198
Gene Name
transmembrane protein 198
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005209.2 6314 Intron NP_001005209.1
NM_001303098.1 6314 Intron NP_001290027.1
XM_017003367.1 6314 Intron XP_016858856.1

View Full Product Details