Product Details

SNP ID
rs3737251
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:32817982 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
AGCGCCGTCTGCAGTCCCCTGCCCA[C/G]TGGGGGTGGGGGCCGCCTCTGTCCC
Phenotype
MIM: 611889 MIM: 603623
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
S100PBP PubMed Links

Gene Details

Gene
S100PBP
Gene Name
S100P binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256121.1 51 Intron NP_001243050.1
NM_022753.3 51 Intron NP_073590.2
XM_011541961.2 51 Intron XP_011540263.1
XM_011541962.2 51 Intron XP_011540264.1
XM_011541963.1 51 Intron XP_011540265.1
XM_017002098.1 51 Intron XP_016857587.1
Gene
YARS
Gene Name
tyrosyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003680.3 51 UTR 5 NP_003671.1
XM_011542347.1 51 Intron XP_011540649.1
XM_011542348.1 51 Intron XP_011540650.1
XM_017002651.1 51 Intron XP_016858140.1

View Full Product Details