Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001141945.2 | Intron | NP_001135417.1 | |||
NM_001320855.1 | Intron | NP_001307784.1 | |||
NM_001613.2 | Intron | NP_001604.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000043.5 | Intron | NP_000034.1 | |||
NM_001320619.1 | Intron | NP_001307548.1 | |||
NM_152871.3 | Intron | NP_690610.1 | |||
NM_152872.3 | Intron | NP_690611.1 | |||
XM_006717819.3 | Intron | XP_006717882.1 | |||
XM_011539764.2 | Intron | XP_011538066.1 | |||
XM_011539765.2 | Intron | XP_011538067.1 | |||
XM_011539766.2 | Intron | XP_011538068.1 | |||
XM_011539767.2 | Intron | XP_011538069.1 |