Product Details
- SNP ID
-
rs3749739
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:38942079 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGTAACTGAAACTCTTAAAACTGTG[A/G]TAATGAATCATGAACCCCTCAAAAG
- Phenotype
-
MIM: 601743
MIM: 609022
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OSMR
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs138291275] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OSMR
- Gene Name
- oncostatin M receptor
- Gene
- RICTOR
- Gene Name
- RPTOR independent companion of MTOR complex 2
View Full Product Details