Product Details

SNP ID
rs3750161
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:47277448 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATGGTGGGGTGGCATGGGGTCTC[C/T]AACGTGGCCAATTCTTCCCACCACC
Phenotype
MIM: 606825
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TNS3 PubMed Links

Gene Details

Gene
TNS3
Gene Name
tensin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022748.11 4884 UTR 3 NP_073585.8
XM_011515476.2 4884 UTR 3 XP_011513778.1
XM_011515477.2 4884 UTR 3 XP_011513779.1
XM_011515478.2 4884 UTR 3 XP_011513780.1
XM_011515479.2 4884 UTR 3 XP_011513781.1
XM_011515480.2 4884 UTR 3 XP_011513782.1
XM_011515481.2 4884 UTR 3 XP_011513783.1
XM_011515482.2 4884 UTR 3 XP_011513784.1
XM_011515483.2 4884 UTR 3 XP_011513785.1
XM_011515484.2 4884 UTR 3 XP_011513786.1
XM_017012537.1 4884 UTR 3 XP_016868026.1
XM_017012538.1 4884 UTR 3 XP_016868027.1
XM_017012539.1 4884 UTR 3 XP_016868028.1

View Full Product Details