Product Details

SNP ID
rs3829814
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:140726457 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTAACAAATTGTACGAACACAAG[A/G]CTTAAGAAATAAGAGCAGATGCTGC
Phenotype
MIM: 164757
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BRAF PubMed Links

Gene Details

Gene
BRAF
Gene Name
B-Raf proto-oncogene, serine/threonine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004333.4 2315 Intron NP_004324.2
XM_005250045.2 2315 UTR 3 XP_005250102.1
XM_017012558.1 2315 UTR 3 XP_016868047.1
XM_017012559.1 2315 Silent Mutation AGC,AGT S769S XP_016868048.1

View Full Product Details