Product Details

SNP ID
rs3786658
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14394529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACAGGAGGCCACTTTGGAAGCTC[A/G]GAACATCCCCCTGCCCATCTGGCAT
Phenotype
MIM: 601211
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADGRE5 PubMed Links

Gene Details

Gene
ADGRE5
Gene Name
adhesion G protein-coupled receptor E5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025160.2 Intron NP_001020331.1
NM_001784.4 Intron NP_001775.2
NM_078481.3 Intron NP_510966.1
XM_011528451.2 Intron XP_011526753.1
XM_017027547.1 Intron XP_016883036.1

View Full Product Details