Product Details

SNP ID
rs3204801
Assay Type
Functionally Tested
NCBI dbSNP Submissions
3
Location
Chr.1:12860049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGGGGAGCTGGAAATTCACAACA[C/T]GTGCTGGCCACATCTGATAAGAAAG
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PRAMEF2 PubMed Links
Additional Information
For this assay, SNP(s) [rs77138000] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
PRAMEF2
Gene Name
PRAME family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023014.1 Intron NP_075390.1
XM_011542004.1 Intron XP_011540306.1

View Full Product Details