Product Details

SNP ID
rs3793018
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43084426 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAAGTGTTGGGATTACAGGCGTA[A/G]GCCATCACGCCCAGCCTGGAAACTG
Phenotype
MIM: 601890
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLC4 PubMed Links

Gene Details

Gene
KLC4
Gene Name
kinesin light chain 4
There are no transcripts associated with this gene.

Gene
PTK7
Gene Name
protein tyrosine kinase 7 (inactive)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270398.1 Intron NP_001257327.1
NM_002821.4 Intron NP_002812.2
NM_152880.3 Intron NP_690619.1
NM_152881.3 Intron NP_690620.1
NM_152882.3 Intron NP_690621.1
XM_011514765.2 Intron XP_011513067.1
XM_011514766.2 Intron XP_011513068.1

View Full Product Details