Product Details
- SNP ID
-
rs3795717
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
7
- Location
-
Chr.1:44810703 on Build GRCh38
- Set Membership
-
HapMap
JSNP
- Context Sequence [VIC/FAM]
- CACACACTCTGGCCTGGAGAGGAAC[A/G]TGTGCCCACACAGAGAGAAGTATGT
- Phenotype
-
MIM: 602913
MIM: 603673
MIM: 611713
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BTBD19
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116724108] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BTBD19
- Gene Name
- BTB domain containing 19
- Gene
- PLK3
- Gene Name
- polo like kinase 3
There are no transcripts associated with this gene.
- Gene
- PTCH2
- Gene Name
- patched 2
There are no transcripts associated with this gene.
- Gene
- TCTEX1D4
- Gene Name
- Tctex1 domain containing 4
View Full Product Details