Product Details

SNP ID
rs3803433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:100881468 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGTTGTACAGGATTTCAAAATACT[A/G]TATATAGAAACCAGACAGTAAATGT
Phenotype
MIM: 600463
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALDH1A3 PubMed Links

Gene Details

Gene
ALDH1A3
Gene Name
aldehyde dehydrogenase 1 family member A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000693.3 Intron NP_000684.2
NM_001293815.1 Intron NP_001280744.1
Gene
LOC105369201
Gene Name
uncharacterized LOC105369201
There are no transcripts associated with this gene.

View Full Product Details