Product Details

SNP ID
rs3825559
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:101732259 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGTTGTTCTAGAGATTGTCAGAC[C/T]GAGACCACGCCACACACCCAGAAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00239 PubMed Links

Gene Details

Gene
LINC00239
Gene Name
long intergenic non-protein coding RNA 239
There are no transcripts associated with this gene.

View Full Product Details