Product Details

SNP ID
rs3946473
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:128162429 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACAAGTTTTCTTGTTTTGCCTGG[G/T]ACCTTATTGCTCTCTGACCCCACCT
Phenotype
MIM: 607695 MIM: 603449
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EEFSEC PubMed Links

Gene Details

Gene
EEFSEC
Gene Name
eukaryotic elongation factor, selenocysteine-tRNA specific
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021937.4 Intron NP_068756.2
Gene
RUVBL1
Gene Name
RuvB like AAA ATPase 1
There are no transcripts associated with this gene.

View Full Product Details