Product Details

SNP ID
rs3735249
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:16792483 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGTTGATTAAGCTCGTGTCTAC[C/T]TACAGTCATCTAGTGAGAACCTGTG
Phenotype
MIM: 606358 MIM: 613139
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AGR2 PubMed Links

Gene Details

Gene
AGR2
Gene Name
anterior gradient 2, protein disulphide isomerase family member
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006408.3 1109 Intron NP_006399.1
XM_005249581.4 1109 UTR 3 XP_005249638.1
Gene
TSPAN13
Gene Name
tetraspanin 13
There are no transcripts associated with this gene.

View Full Product Details