Product Details
- SNP ID
-
rs3745205
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:40442295 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTAAGAAAATAAGATCTGGCGTTA[A/C]GTACAGCTAGATTTAAATCTCACTT
- Phenotype
-
MIM: 600941
MIM: 612125
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
BLVRB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139647276] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BLVRB
- Gene Name
- biliverdin reductase B
There are no transcripts associated with this gene.
- Gene
- LOC105372489
- Gene Name
- uncharacterized LOC105372489
There are no transcripts associated with this gene.
- Gene
- SERTAD3
- Gene Name
- SERTA domain containing 3
View Full Product Details