Product Details

SNP ID
rs3747006
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:46301747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGAGGGGTCCCTGGGAGGGGCTG[C/T]TGCCCTGGTGGGGTTCACAGGCCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C21orf58 PubMed Links

Gene Details

Gene
C21orf58
Gene Name
chromosome 21 open reading frame 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286462.1 4548 Intron NP_001273391.1
NM_001286463.1 4548 UTR 3 NP_001273392.1
NM_001286476.1 4548 Intron NP_001273405.1
NM_001286477.1 4548 UTR 3 NP_001273406.1
NM_058180.4 4548 UTR 3 NP_478060.2
XM_005261148.4 4548 Intron XP_005261205.1
XM_006724018.3 4548 Intron XP_006724081.1
XM_011529619.2 4548 Intron XP_011527921.1
XM_011529620.2 4548 Intron XP_011527922.1
XM_011529621.2 4548 UTR 3 XP_011527923.1
XM_011529623.2 4548 UTR 3 XP_011527925.1
XM_011529624.2 4548 UTR 3 XP_011527926.1
XM_011529625.2 4548 UTR 3 XP_011527927.1
XM_011529626.2 4548 UTR 3 XP_011527928.1
XM_011529627.2 4548 UTR 3 XP_011527929.1
XM_011529628.1 4548 Intron XP_011527930.1
XM_017028388.1 4548 UTR 3 XP_016883877.1
XM_017028389.1 4548 Intron XP_016883878.1
XM_017028390.1 4548 UTR 3 XP_016883879.1
XM_017028391.1 4548 UTR 3 XP_016883880.1
XM_017028392.1 4548 UTR 3 XP_016883881.1
XM_017028393.1 4548 UTR 3 XP_016883882.1
Gene
YBEY
Gene Name
ybeY metallopeptidase (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006114.2 4548 Intron NP_001006114.1
NM_001314022.1 4548 Intron NP_001300951.1
NM_001314023.1 4548 Intron NP_001300952.1
NM_001314024.1 4548 Intron NP_001300953.1
NM_001314025.1 4548 Intron NP_001300954.1
NM_001314026.1 4548 Intron NP_001300955.1
NM_058181.2 4548 Intron NP_478061.1
XM_005261156.3 4548 Intron XP_005261213.2
XM_011529629.2 4548 Intron XP_011527931.1
XM_011529630.2 4548 Intron XP_011527932.1
XM_011529631.2 4548 Intron XP_011527933.1
XM_011529633.2 4548 Intron XP_011527935.1
XM_011529634.2 4548 Intron XP_011527936.2
XM_011529635.2 4548 Intron XP_011527937.1
XM_017028394.1 4548 Intron XP_016883883.1
XM_017028395.1 4548 Intron XP_016883884.1
XM_017028396.1 4548 Intron XP_016883885.1

View Full Product Details