Product Details

SNP ID
rs3748209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:135562960 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTAGCCACGCTCTCCCAGAGGCG[A/G]CTCTGCTGGGGCATGAGGGAGTGGG
Phenotype
MIM: 173310
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PAEP PubMed Links

Gene Details

Gene
PAEP
Gene Name
progestagen associated endometrial protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018048.1 Intron NP_001018058.1
NM_001018049.2 Intron NP_001018059.1
NM_002571.3 Intron NP_002562.2
XM_011518745.2 Intron XP_011517047.1
XM_011518746.2 Intron XP_011517048.1
XM_011518747.2 Intron XP_011517049.1
XM_011518748.1 Intron XP_011517050.1
XM_011518749.2 Intron XP_011517051.1
XM_011518751.1 Intron XP_011517053.1
XM_011518752.2 Intron XP_011517054.1
XM_017014782.1 Intron XP_016870271.1
XM_017014783.1 Intron XP_016870272.1

View Full Product Details