Product Details

SNP ID
rs3749477
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:177431392 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGATGATTCCTTTTGGGTTTAATA[C/T]ATCACTGTGGAAATAAATCTCAAAG
Phenotype
MIM: 613228
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AGA PubMed Links

Gene Details

Gene
AGA
Gene Name
aspartylglucosaminidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000027.3 1455 UTR 3 NP_000018.2
NM_001171988.1 1455 UTR 3 NP_001165459.1

View Full Product Details