Product Details

SNP ID
rs3754230
Assay Type
Functionally tested
NCBI dbSNP Submissions
57
Location
Chr.1:235663283 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GACAAGTTGAGATGCTCATAAATGC[A/G]GTGATATAGAATAAAATCAGGATCA
Phenotype
MIM: 606897
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LYST PubMed Links

Gene Details

Gene
LYST
Gene Name
lysosomal trafficking regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000081.3 Intron NP_000072.2
NM_001301365.1 Intron NP_001288294.1
XM_011544031.1 Intron XP_011542333.1
XM_011544032.1 Intron XP_011542334.1
XM_011544033.2 Intron XP_011542335.1
XM_011544034.1 Intron XP_011542336.1
XM_011544035.2 Intron XP_011542337.1
XM_011544036.2 Intron XP_011542338.1
XM_011544037.2 Intron XP_011542339.1
XM_011544039.2 Intron XP_011542341.1
XM_011544040.2 Intron XP_011542342.1
XM_017000150.1 Intron XP_016855639.1
XM_017000151.1 Intron XP_016855640.1

View Full Product Details