Product Details

SNP ID
rs3766744
Assay Type
Functionally tested
NCBI dbSNP Submissions
40
Location
Chr.1:11983660 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
TCAAGGCTTTCCAGAGGCAGAGGAG[A/G]AACCCAGCTGCCCTGCATCACCCTG
Phenotype
MIM: 608507 MIM: 153454
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MFN2 PubMed Links

Gene Details

Gene
MFN2
Gene Name
mitofusin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127660.1 Intron NP_001121132.1
NM_014874.3 Intron NP_055689.1
XM_005263543.3 Intron XP_005263600.1
XM_005263545.3 Intron XP_005263602.1
XM_005263547.3 Intron XP_005263604.1
XM_005263548.3 Intron XP_005263605.1
Gene
PLOD1
Gene Name
procollagen-lysine,2-oxoglutarate 5-dioxygenase 1
There are no transcripts associated with this gene.

View Full Product Details