Product Details

SNP ID
rs3788336
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23060802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCACTGCCTCCCAGGGCCTGGCAC[A/G]TGGCAGGGGCTTCAGGACCCTCAGC
Phenotype
MIM: 139160 MIM: 605663
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNAZ PubMed Links

Gene Details

Gene
GNAZ
Gene Name
G protein subunit alpha z
There are no transcripts associated with this gene.

Gene
RSPH14
Gene Name
radial spoke head 14 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014433.2 Intron NP_055248.1
XM_011530149.2 Intron XP_011528451.1
XM_011530150.1 Intron XP_011528452.1
XM_011530151.2 Intron XP_011528453.1
XM_011530152.2 Intron XP_011528454.1
XM_011530154.2 Intron XP_011528456.1
XM_011530155.2 Intron XP_011528457.1
XM_017028774.1 Intron XP_016884263.1

View Full Product Details