Product Details

SNP ID
rs3793659
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136667383 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGACCCTGCAGCATGGCAGGGGCTC[A/G]CCAGGTCTCCAGACCCCTGGCTGCG
Phenotype
MIM: 603100 MIM: 608582 MIM: 611767
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AGPAT2 PubMed Links

Gene Details

Gene
AGPAT2
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 2
There are no transcripts associated with this gene.

Gene
EGFL7
Gene Name
EGF like domain multiple 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016215.4 Intron NP_057299.1
NM_201446.2 Intron NP_958854.1
XM_006717141.3 Intron XP_006717204.1
XM_011518764.1 Intron XP_011517066.1
XM_011518765.1 Intron XP_011517067.1
XM_011518766.1 Intron XP_011517068.1
XM_011518767.1 Intron XP_011517069.1
XM_011518768.1 Intron XP_011517070.1
XM_017014795.1 Intron XP_016870284.1
XM_017014796.1 Intron XP_016870285.1
XM_017014797.1 Intron XP_016870286.1
XM_017014798.1 Intron XP_016870287.1
XM_017014799.1 Intron XP_016870288.1
Gene
HSPC324
Gene Name
uncharacterized LOC101928612
There are no transcripts associated with this gene.

Gene
MIR126
Gene Name
microRNA 126
There are no transcripts associated with this gene.

View Full Product Details