Product Details

SNP ID
rs3794416
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:113159636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGTTAAATGCTGCGCACAGAGGC[A/G]GCACAGGAGCTGATGGCTCTTGGTC
Phenotype
MIM: 613713 MIM: 176895
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC102724474 PubMed Links

Gene Details

Gene
LOC102724474
Gene Name
WAS/WASL-interacting protein family member 2-like
There are no transcripts associated with this gene.

Gene
PCID2
Gene Name
PCI domain containing 2
There are no transcripts associated with this gene.

Gene
PROZ
Gene Name
protein Z, vitamin K dependent plasma glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256134.1 Intron NP_001243063.1
NM_003891.2 Intron NP_003882.1
XM_017020812.1 Intron XP_016876301.1
XM_017020813.1 Intron XP_016876302.1

View Full Product Details