Product Details

SNP ID
rs3794638
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89687028 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGGGGCGGGCTCAGGGATGCCTCG[A/C]GCCCGCGGAGAGACGGGCCCGGGAC
Phenotype
MIM: 603464
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CDK10 PubMed Links

Gene Details

Gene
CDK10
Gene Name
cyclin dependent kinase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098533.2 224 Intron NP_001092003.2
NM_001160367.1 224 Intron NP_001153839.1
NM_052987.3 224 Intron NP_443713.2
NM_052988.4 224 Intron NP_443714.3
XM_006721308.2 224 Intron XP_006721371.1
XM_006721310.2 224 Intron XP_006721373.1
XM_011523405.2 224 Intron XP_011521707.1
XM_011523406.2 224 Intron XP_011521708.1
XM_011523407.2 224 Intron XP_011521709.1
XM_011523408.2 224 Intron XP_011521710.1
XM_011523410.2 224 Intron XP_011521712.1
XM_011523411.2 224 Intron XP_011521713.1
XM_011523412.1 224 Intron XP_011521714.1
XM_011523413.2 224 UTR 5 XP_011521715.1
XM_011523414.2 224 UTR 5 XP_011521716.1
XM_011523415.2 224 Intron XP_011521717.1
XM_011523416.2 224 Intron XP_011521718.1
XM_011523417.2 224 Intron XP_011521719.1
XM_017023806.1 224 Intron XP_016879295.1
XM_017023807.1 224 Intron XP_016879296.1
XM_017023808.1 224 Intron XP_016879297.1
XM_017023809.1 224 Intron XP_016879298.1
XM_017023810.1 224 UTR 5 XP_016879299.1
Gene
SPATA2L
Gene Name
spermatogenesis associated 2 like
There are no transcripts associated with this gene.

View Full Product Details