Product Details

SNP ID
rs3797876
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:80629265 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAATAGTGCATAAAGCAAATAAAA[A/G]GTTTCCCTTTTTTCTTCACAATGGT
Phenotype
MIM: 126060
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHFR PubMed Links

Gene Details

Gene
DHFR
Gene Name
dihydrofolate reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000791.3 Intron NP_000782.1
NM_001290354.1 Intron NP_001277283.1
NM_001290357.1 Intron NP_001277286.1
Gene
LINC01337
Gene Name
long intergenic non-protein coding RNA 1337
There are no transcripts associated with this gene.

View Full Product Details