Product Details

SNP ID
rs3850021
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:103969201 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACCAATTATTAGGTACTATTCCC[A/T]TTTTACAGATGTGGAAGCATGGGCA
Phenotype
MIM: 601423
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C12orf73 PubMed Links

Gene Details

Gene
C12orf73
Gene Name
chromosome 12 open reading frame 73
There are no transcripts associated with this gene.

Gene
TDG
Gene Name
thymine DNA glycosylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003211.4 Intron NP_003202.3
XM_005269125.1 Intron XP_005269182.1
XM_011538714.1 Intron XP_011537016.1
XM_011538715.1 Intron XP_011537017.1

View Full Product Details