Product Details

SNP ID
rs3807123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:130021547 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCAATGGCTAGCAGAGTCACAGT[A/C]TGGGCTGTGGAACAGAGTAAAGTAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZC3HC1 PubMed Links

Gene Details

Gene
ZC3HC1
Gene Name
zinc finger C3HC-type containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282190.1 Intron NP_001269119.1
NM_001282191.1 Intron NP_001269120.1
NM_016478.4 Intron NP_057562.3
XM_005250403.2 Intron XP_005250460.1
XM_011516288.2 Intron XP_011514590.1
XM_011516289.2 Intron XP_011514591.1
XM_011516290.2 Intron XP_011514592.1
XM_017012287.1 Intron XP_016867776.1
XM_017012288.1 Intron XP_016867777.1

View Full Product Details