Product Details

SNP ID
rs3809793
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:29591038 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGGTGTGTAGGGGGGAGCACCAC[C/T]TGCCGCCCCGAGCTGCTCTCTCACC
Phenotype
MIM: 615124 MIM: 608434
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ANKRD13B PubMed Links

Gene Details

Gene
ANKRD13B
Gene Name
ankyrin repeat domain 13B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152345.4 Intron NP_689558.4
XM_005257920.4 Intron XP_005257977.1
XM_011524319.2 Intron XP_011522621.1
XM_011524320.2 Intron XP_011522622.1
XM_011524321.2 Intron XP_011522623.1
XM_017024174.1 Intron XP_016879663.1
XM_017024175.1 Intron XP_016879664.1
XM_017024176.1 Intron XP_016879665.1
Gene
GIT1
Gene Name
GIT ArfGAP 1
There are no transcripts associated with this gene.

View Full Product Details