Product Details

SNP ID
rs3943574
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18732176 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGTGGACAGATTGGGCACTACAGG[C/T]CAGAAGCCCAGGATCCCAGGTGAAG
Phenotype
MIM: 176879
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGSF22 PubMed Links

Gene Details

Gene
IGSF22
Gene Name
immunoglobulin superfamily member 22
There are no transcripts associated with this gene.

Gene
PTPN5
Gene Name
protein tyrosine phosphatase, non-receptor type 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039970.1 2115 Intron NP_001035059.1
NM_001278236.1 2115 Intron NP_001265165.1
NM_001278238.1 2115 Intron NP_001265167.1
NM_001278239.1 2115 Intron NP_001265168.1
NM_006906.1 2115 Intron NP_008837.1
NM_032781.3 2115 Intron NP_116170.3
XM_011520411.2 2115 Intron XP_011518713.1
XM_017018434.1 2115 Intron XP_016873923.1
XM_017018435.1 2115 Intron XP_016873924.1
XM_017018436.1 2115 Intron XP_016873925.1
XM_017018437.1 2115 Intron XP_016873926.1
XM_017018438.1 2115 Intron XP_016873927.1
XM_017018439.1 2115 Intron XP_016873928.1
XM_017018440.1 2115 Intron XP_016873929.1
XM_017018441.1 2115 UTR 3 XP_016873930.1

View Full Product Details