Product Details

SNP ID
rs3819021
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:21008417 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTTACAATGTTTCTTAAAATGTT[A/G]TCTCAAAATACATTTGTCCACCTAG
Phenotype
MIM: 615941
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HACD4 PubMed Links

Gene Details

Gene
HACD4
Gene Name
3-hydroxyacyl-CoA dehydratase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010915.4 Intron NP_001010915.2
NM_001321883.1 Intron NP_001308812.1
NM_001321903.1 Intron NP_001308832.1
XM_011517876.2 Intron XP_011516178.1
XM_017014713.1 Intron XP_016870202.1
XM_017014714.1 Intron XP_016870203.1

View Full Product Details