Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001017395.3 | 2097 | Missense Mutation | NP_001017395.2 | ||
NM_001128224.2 | 2097 | Missense Mutation | NP_001121696.1 | ||
XM_006713542.3 | 2097 | Missense Mutation | XP_006713605.1 | ||
XM_006713543.3 | 2097 | Missense Mutation | XP_006713606.1 | ||
XM_006713544.1 | 2097 | Missense Mutation | XP_006713607.1 | ||
XM_006713550.3 | 2097 | Missense Mutation | XP_006713613.1 | ||
XM_006713552.3 | 2097 | Missense Mutation | XP_006713615.1 | ||
XM_006713553.3 | 2097 | Missense Mutation | XP_006713616.1 | ||
XM_011512572.2 | 2097 | Missense Mutation | XP_011510874.1 | ||
XM_011512573.2 | 2097 | Missense Mutation | XP_011510875.1 | ||
XM_011512576.2 | 2097 | Missense Mutation | XP_011510878.1 | ||
XM_011512579.2 | 2097 | Missense Mutation | XP_011510881.1 | ||
XM_011512580.2 | 2097 | Missense Mutation | XP_011510882.1 | ||
XM_011512581.2 | 2097 | Missense Mutation | XP_011510883.1 | ||
XM_011512582.1 | 2097 | Missense Mutation | XP_011510884.1 | ||
XM_011512583.2 | 2097 | Missense Mutation | XP_011510885.1 | ||
XM_011512584.2 | 2097 | Missense Mutation | XP_011510886.1 | ||
XM_017005931.1 | 2097 | Missense Mutation | XP_016861420.1 | ||
XM_017005932.1 | 2097 | Missense Mutation | XP_016861421.1 | ||
XM_017005933.1 | 2097 | Missense Mutation | XP_016861422.1 | ||
XM_017005934.1 | 2097 | Missense Mutation | XP_016861423.1 | ||
XM_017005935.1 | 2097 | Missense Mutation | XP_016861424.1 | ||
XM_017005936.1 | 2097 | Missense Mutation | XP_016861425.1 | ||
XM_017005937.1 | 2097 | Missense Mutation | XP_016861426.1 | ||
XM_017005938.1 | 2097 | Missense Mutation | XP_016861427.1 | ||
XM_017005939.1 | 2097 | Missense Mutation | XP_016861428.1 | ||
XM_017005940.1 | 2097 | Missense Mutation | XP_016861429.1 | ||
XM_017005941.1 | 2097 | Missense Mutation | XP_016861430.1 |