Product Details

SNP ID
rs3826541
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48621594 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCAGGCCCATGAAGCAGGAGGC[A/C]AAGCCCGGCCCTGGGGCGGGTCTCC
Phenotype
MIM: 142963 MIM: 142964
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HOXB8 PubMed Links

Gene Details

Gene
HOXB8
Gene Name
homeobox B8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024016.3 2263 Intron NP_076921.1
XM_005257286.2 2263 UTR 5 XP_005257343.1
XM_017024564.1 2263 UTR 5 XP_016880053.1
Gene
HOXB9
Gene Name
homeobox B9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024017.4 2263 UTR 3 NP_076922.1

View Full Product Details