Product Details

SNP ID
rs61753185
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:89178183 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCCTTCACAGGGGTGGATGACC[A/G]GGAGTCGTGGCCTTCCGTCTTTTAT
Phenotype
MIM: 606933
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TYR PubMed Links

Gene Details

Gene
TYR
Gene Name
tyrosinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000372.4 619 Missense Mutation CAG,CGG Q77R NP_000363.1
XM_011542970.2 619 Missense Mutation CAG,CGG Q77R XP_011541272.1

View Full Product Details