Product Details
- SNP ID
-
rs2464171
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:91051183 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AATTAATGCAGGCTGTGATGCATGT[A/G]ACTGGCAAAAGCATCTTTGAATAGA
- Phenotype
-
MIM: 603288
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KERA
PubMed Links
Gene Details
- Gene
- KERA
- Gene Name
- keratocan
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007035.3 |
1841 |
UTR 3 |
|
|
NP_008966.1 |
View Full Product Details