Product Details
- SNP ID
-
hCV27541080
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:85473735 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGATGGCACTTACAGATTGATTC[A/C]GTTTCACTTTCACTGGGGTTCACTT
- Phenotype
-
MIM: 611492
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CA2
PubMed Links
Gene Details
- Gene
- CA2
- Gene Name
- carbonic anhydrase 2
- Gene
- CA3-AS1
- Gene Name
- CA3 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details