Product Details

SNP ID
rs122462166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:137567495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGAGCCGGCCCAAGAAGAGCTG[A/C]GACCGGACCTTCAGCACCATGCATG
Phenotype
MIM: 300265
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZIC3 PubMed Links

Gene Details

Gene
ZIC3
Gene Name
Zic family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003413.3 1309 Nonsense Mutation TGA,TGC *268C NP_003404.1
XM_017029802.1 1309 Nonsense Mutation TGA,TGC *268C XP_016885291.1

View Full Product Details