Product Details
- SNP ID
-
rs78761037
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:38105428 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCTGGCCTTAAAGCGGTCGTGGAA[C/T]TATGCTTCCTCAGGCAGGCGGTGGA
- Phenotype
-
MIM: 604782
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ASH2L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75541171] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ASH2L
- Gene Name
- ASH2 like histone lysine methyltransferase complex subunit
View Full Product Details