Product Details

SNP ID
rs79945090
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:112074935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAGGGGTGAGTGAGCGTTGGGGT[A/G]GTTGCACAAAGCTCCTGCTAGCTAC
Phenotype
MIM: 606659
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C11orf57 PubMed Links

Gene Details

Gene
C11orf57
Gene Name
chromosome 11 open reading frame 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001082969.1 Intron NP_001076438.1
NM_001082970.1 Intron NP_001076439.1
NM_001301017.1 Intron NP_001287946.1
NM_001301019.1 Intron NP_001287948.1
NM_001301021.1 Intron NP_001287950.1
NM_018195.3 Intron NP_060665.3
Gene
PIH1D2
Gene Name
PIH1 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001082619.1 Intron NP_001076088.1
NM_138789.3 Intron NP_620144.1
XM_011542593.2 Intron XP_011540895.2
XM_017017201.1 Intron XP_016872690.1
XM_017017202.1 Intron XP_016872691.1
XM_017017203.1 Intron XP_016872692.1
XM_017017204.1 Intron XP_016872693.1
XM_017017205.1 Intron XP_016872694.1
Gene
TIMM8B
Gene Name
translocase of inner mitochondrial membrane 8 homolog B
There are no transcripts associated with this gene.

View Full Product Details