Product Details

SNP ID
rs141572324
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:108716508 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGAGATTTTCCTCCAGCATATA[C/T]AAGTGTCATAATTTTCTTTCAATGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FNDC7 PubMed Links

Gene Details

Gene
FNDC7
Gene Name
fibronectin type III domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144937.1 Intron NP_001138409.1

View Full Product Details