Product Details

SNP ID
rs374928393
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:82363832 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTGGGGTGCGAAGAGAGTCGGC[C/G]CCCGCAACGCGGAGCCGGGAAGTCG
Phenotype
MIM: 190920
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
TPBG PubMed Links

Gene Details

Gene
TPBG
Gene Name
trophoblast glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166392.1 521 Intron NP_001159864.1
NM_006670.4 521 UTR 5 NP_006661.1
XM_011536097.2 521 UTR 5 XP_011534399.1

View Full Product Details