Product Details

SNP ID
rs45571738
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43353914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCATGTGTGGAAGGTGTCCGACCT[A/G]CCCCGGCAATGGACCCCTAAGAACA
Phenotype
MIM: 162860 MIM: 605157
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD177 PubMed Links

Gene Details

Gene
CD177
Gene Name
CD177 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020406.3 170 Silent Mutation CTA,CTG L38L NP_065139.2
XM_017027021.1 170 Silent Mutation CTA,CTG L38L XP_016882510.1
XM_017027022.1 170 Silent Mutation CTA,CTG L38L XP_016882511.1
Gene
PRG1
Gene Name
p53-responsive gene 1
There are no transcripts associated with this gene.

View Full Product Details