Product Details

SNP ID
rs1033844484
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50719664 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGAGCGGGCCGAGGAGATTGGC[C/G]ACGGTGTCGCCCGTGTTTTCGTTGG
Phenotype
MIM: 609434
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LUC7L3 PubMed Links

Gene Details

Gene
LUC7L3
Gene Name
LUC7 like 3 pre-mRNA splicing factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006107.3 100 UTR 5 NP_006098.2
NM_016424.4 100 UTR 5 NP_057508.2

View Full Product Details