Product Details

SNP ID
rs78892734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18708272 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCACCATGTGACTCTTCAGCCGGG[C/T]ACTGAGGTCAAACTTGGGTGCAGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGSF22 PubMed Links

Gene Details

Gene
IGSF22
Gene Name
immunoglobulin superfamily member 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173588.3 4030 Missense Mutation ACC,GCC T1008A NP_775859.3
XM_011520030.2 4030 Missense Mutation ACC,GCC T1028A XP_011518332.1
XM_011520031.2 4030 Missense Mutation ACC,GCC T927A XP_011518333.1
XM_017017607.1 4030 Missense Mutation ACC,GCC T1028A XP_016873096.1
XM_017017608.1 4030 Missense Mutation ACC,GCC T1008A XP_016873097.1
Gene
TMEM86A
Gene Name
transmembrane protein 86A
There are no transcripts associated with this gene.

View Full Product Details