Product Details

SNP ID
rs145211350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110124792 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACCCAGGCAGCACGGTTTCTCT[C/T]CCAAATTGTGCCAGAGAACACGCAC
Phenotype
MIM: 605489
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFT81 PubMed Links

Gene Details

Gene
IFT81
Gene Name
intraflagellar transport 81
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143779.1 458 Intron NP_001137251.1
NM_014055.3 458 UTR 5 NP_054774.2
NM_031473.3 458 UTR 5 NP_113661.2
XM_011538232.2 458 Intron XP_011536534.1
XM_017019217.1 458 Intron XP_016874706.1
XM_017019218.1 458 UTR 5 XP_016874707.1

View Full Product Details