Product Details

SNP ID
rs1045067585
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:54920812 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGGACCACTTCCGGCCAGGGAG[A/G]GGGATAGGCTCAGACACAAGGAGAG
Phenotype
MIM: 300132
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRO PubMed Links

Gene Details

Gene
TRO
Gene Name
trophinin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039705.2 250 UTR 5 NP_001034794.1
NM_001271183.1 250 UTR 5 NP_001258112.1
NM_001271184.1 250 Intron NP_001258113.1
NM_016157.3 250 UTR 5 NP_057241.2
NM_177556.2 250 UTR 5 NP_808224.1
NM_177557.2 250 UTR 5 NP_808225.1

View Full Product Details